Skip to Content
Merck
All Photos(1)

Key Documents

DX1726

Supelco

N,N-Dimethylformamide

OmniSolv®

Synonym(s):

Formic Acid Dimethylamide, DMF, DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-679-5
Assay:
≥99.9% (GC)
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.5 (vs air)

Quality Level

vapor pressure

2.7 mmHg ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

833 °F

shelf life

24 mo.

expl. lim.

15.2 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

<0.05% Water

evapn. residue

<5 ppm

color

APHA: 10

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L in H2O)

bp

153 °C (lit.)

mp

−61 °C (lit.)

transition temp

flash point 57.5 °C

density

0.944 g/mL (lit.)

UV cutoff

≤268 nm

UV absorption

λ: 268 nm Amax: ≤1.00
λ: 270 nm Amax: ≤0.75
λ: 275 nm Amax: ≤0.30
λ: 295 nm Amax: ≤0.07
λ: 310 nm Amax: ≤0.03
λ: 340 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DX1726P-1 is packaged in poly coated glass bottles
OmniSolv®
Solvent for many hydrophobic organic compounds.

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC): 99.9% min
Color (APHA): 10 max
Evaporation residue: 5ppm max
Expiration date: 24 months
Filtered through 0.2 m filter: To pass test
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Titrable acid: eq/g
Titrable base: eq/g
UV Abs. at 268 nm: 1.00AU max
UV Abs. at 270 nm: 0.75AU max
UV Abs. at 275 nm: 0.30AU max
UV Abs. at 295 nm: 0.07AU max
UV Abs. at 310 nm: 0.03AU max
UV Abs. at 340 nm: 0.01AU max
UV Cut-off: 268nm max
Water (H2O): 0.05% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service