Skip to Content
Merck
All Photos(1)

Key Documents

DX0657

Sigma-Aldrich

Dichloroacetic Acid

Biotechnology

Synonym(s):

Dichloroacetic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2Cl2O2
CAS Number:
Molecular Weight:
128.94
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-207-0
Pricing and availability is not currently available.

vapor pressure

0.19 hPa ( 20 °C)

Quality Level

form

liquid

pH

1.2 (20 °C, 129 g/L in H2O)

bp

192 °C/1013 hPa (decomposes)

mp

-4 °C

shipped in

ambient

storage temp.

room temp

SMILES string

ClC(Cl)C(=O)O

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

JXTHNDFMNIQAHM-UHFFFAOYSA-N

Application

Biotechnology

Analysis Note

Absorbance at 310 nmReport onlyAUAssay (titration)99% minFormClear liquidFreezing point12°C minIdentity (IR-spectrum)Monochloroacetic acid0.20% maxTrichloroacetic acid1.00% maxWater (H2O)1000ppm max

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Lact. - Met. Corr. 1 - Repr. 1B - Skin Corr. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Brain,Liver,Testes, Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service