Skip to Content
Merck
All Photos(1)

Key Documents

CBL479

Sigma-Aldrich

Anti-Integrin β3 Antibody, clone PM6/13

clone PM6/13, Chemicon®, from mouse

Synonym(s):

CD61

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

PM6/13, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB3(3690)

Specificity

The antibody is specific for the 110 kDa platelet glycoprotein GP IIIa and inhibits platelet aggregation and activation induced by thrombin and collagen. The platelet glycoprotein is the integrin beta3 chain of the vitronectin receptor and GPIIb/IIIa. The antigen is present on platelets and megakaryocytes.

Antigen distribution:

Platelets >98%

Megakaryocytes >98%

Granulocytes 0%

Monocytes 0%

LGL cells 0%

B cells 0%

T cells 0%

Peripheral blood lymphocytes 0%

FUSION PARTNER: SP2/0 myeloma cell line

Immunogen

Human platelet plasma membrane

Application

FACS: As a marker for megakaryoblastic/cytic leukaemias and idiopathic thrombocythemia





Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Integrin β3 Antibody, clone PM6/13 is validated for use in FC, IP, WB, IH, IH(P) for the detection of Integrin β3.

Linkage

Replaces: MAB1381

Physical form

Format: Purified
The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Boettiger et al.
Molecular biology of the cell, 12(5), 1227-1237 (2001-05-22)
Integrin receptors serve as mechanical links between the cell and its structural environment. Using alpha(v)beta3 integrin expressed in K562 cells as a model system, the process by which the mechanical connection between alpha(v)beta3 and vitronectin develops was analyzed by measuring
Pharmacology of the novel antiangiogenic peptide ATN-161 (Ac-PHSCN-NH2): observation of a U-shaped dose-response curve in several preclinical models of angiogenesis and tumor growth.
Do?ate, F; Parry, GC; Shaked, Y; Hensley, H; Guan, X; Beck, I; Tel-Tsur, Z; Plunkett et al.
Clinical cancer research : an official journal of the American Association for Cancer Research null
D Ma et al.
Thrombosis and haemostasis, 105(6), 1032-1045 (2011-04-09)
A novel family of RGD-containing molecules (Tablysin-15) has been molecularly characterised from the salivary gland of the haematophagous horsefly Tabanus yao. Tablysin-15 does not share primary sequence homology to any disintegrin discovered so far, and displays an RGD motif in
Vineet Gupta et al.
Journal of immunology (Baltimore, Md. : 1950), 180(3), 1713-1718 (2008-01-23)
Formation of the integrin alphabeta heterodimer is essential for cell surface expression and function. At the core of the alphabeta interface is a conserved Arg/Lys "finger" from the beta-subunit that inserts into a cup-like "cage" formed of two layers of
Loss of dipeptidyl peptidase IV immunostaining discriminates malignant melanomas from deep penetrating nevi.
Roesch, A; Wittschier, S; Becker, B; Landthaler, M; Vogt, T
Modern Pathology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service