Skip to Content
Merck
All Photos(1)

Key Documents

CBL440

Sigma-Aldrich

Anti-c-Fos Antibody

CHEMICON®, sheep polyclonal

Synonym(s):

G0/G1 Switch Regulatory Protein 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
WB
species reactivity:
human, rat
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
1

Product Name

Anti-c-Fos Antibody, Chemicon®, from sheep

biological source

sheep

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOS(2353)

Specificity

c-fos is a 55 kDa nuclear associated protein that is rapidly induced in HL60, HeLa and NIH3T3 cells by a variety of agents. c-fos is also induced in neuronal tissue following drug induced seizures.

KNOWN SPECIES CROSS REACTIVITY: Known to cross-react with rat

Immunogen

Synthetic peptide: Met-Phe-Ser-Gly-Phe-Asn-Ala-Asp-Tyr-Glu-Ala-Ser-Ser-Ser-Arg-Cys, selected from a conserved region of mouse and human c-fos.

Application

Anti-c-Fos Antibody is a Sheep Polyclonal Antibody for detection of c-Fos also known as G0/G1 Switch Regulatory Protein 7 & has been validated in WB, ICC.
Immunoblotting: Detergent lysates were produced from HeLa and HL-60 cells stimulated to express c-fos protein. CBL 440 detects a band at 55-60 kDa on immunoblots of stimulated HeLa and HL-60 lysates.

Immunocytochemistry: CBL 440 has been used in immunocytochemistry of vibratome sections of rat brains following either surgical damage to one of the hemispheres (where CBL 440 detects c-fos and c-fos related proteins in the nuclei of neurons in the damaged hemisphere) or kainic acid treatment.

Optimal working dilutions must be determined by the end user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Physical form

Format: Purified
The purified immunoglobulin is presented at a concentration of 100μg/ml in phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Haiyang Xu et al.
Psychopharmacology, 234(16), 2475-2487 (2017-05-27)
The low self-administration (LS)/Kgras (LS) and high self-administration (HS)/Kgras (HS) rat lines were generated by selective breeding for low- and high-intravenous cocaine self-administration, respectively, from a common outbred Wistar stock (Crl:WI). This trait has remained stable after 13 generations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service