Skip to Content
Merck
All Photos(2)

Key Documents

ABN101

Sigma-Aldrich

Anti-GAD65 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Glutamate decarboxylase 2, 65 kDa glutamic acid decarboxylase, GAD-65, Glutamate decarboxylase 65 kDa isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat, pig

species reactivity (predicted by homology)

canine (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GAD2(2572)

General description

GAD65 is a group II decarboxylase enzyme that catalyzes the biosynthesis of GABA from glutamate with the aid of the cofactor, pyridoxal 5′-phosphate. GAD65 localizes at presynaptic GABAergic terminals and is reported to supply local vesicular pools of GABA facilitating transient inhibitiory neurotransmission, although GAD65 may also play a role in more sustained tonic inhibitory signals. It is present as a dimer of alpha and beta subunits and is regulated by alternative splicing, which may produce the GAD67 isoform. GAD65 also undergoes post-translational modifications such as palmitoylation of N-terminal cysteine residues and phosphorylation of multiple alpha-subunit serine residues that enable membrane-anchoring and other vesicular activities. GAD65 has been implicated in neurological conditions resulting from failure of inhibitory neurotransmission such as Huntington′s disease and epilepsy. In addition to its CNS presence, GAD65 also produces GABA in pancreatic beta cells and dysfunction in this area may contribute to the pathogenesis of insulin-dependent diabetes mellitus.

Immunogen

KLH-conjugated linear peptide corresponding to human GAD65.

Application

Anti-GAD Antibody is an antibody against GAD65 for use in Western Blotting. See below for more information about this anti GAD antibody.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
Western Blot Analysis: 1 µg/mL from a representative lot detected GAD65 on 10 µg of rat brain membrane tissue lysate. Western Blot performed without Tween.

Quality

Evaluated by Western Blot in rat brain membrane tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected GAD65 on 10 µg of rat brain membrane tissue lysate.

Target description

~59/63 kDa observed. The calculated molecular weight is 65 kDa, however GAD65 has been shown as a ~59/63 doublet in western blots. (Christgau, S., et al. (1991). Journal of Biological Chemistry. 266(31):21257-21264.) An uncharacterized band appears at ~38 kDa in some lysates.

Physical form

Affinity purified
Purified Rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain membrane tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dany Arsenault et al.
Biology of sex differences, 11(1), 63-63 (2020-11-18)
While the higher prevalence of Alzheimer's disease (AD) in women is clear, studies suggest that biological sex may also influence AD pathogenesis. However, mechanisms behind these differences are not clear. To investigate physiological differences between sexes at the cellular level in the
Iben Lundgaard et al.
PLoS biology, 11(12), e1001743-e1001743 (2014-01-07)
Myelination is essential for rapid impulse conduction in the CNS, but what determines whether an individual axon becomes myelinated remains unknown. Here we show, using a myelinating coculture system, that there are two distinct modes of myelination, one that is
C-Z Yu et al.
European journal of pain (London, England), 18(1), 76-85 (2013-09-17)
B vitamins have been demonstrated to be effective in treating chronic pain due to peripheral nerve injury. We investigated whether B vitamins could alleviate neuropathic pain and reduce neuron injury following temporary ischaemia in a rat model of spinal cord
Wei Li et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(19), 8423-8441 (2013-05-10)
Profound synapse loss is one of the major pathological hallmarks associated with Alzheimer's disease (AD) and might underlie memory impairment. Our previous work demonstrated that the magnesium ion is a critical factor in controlling synapse density/plasticity. Here, we investigated whether
Cyril Bories et al.
Aging, 9(5), 1386-1403 (2017-05-20)
Defects in p21-activated kinase (PAK) lead to dendritic spine abnormalities and are sufficient to cause cognition impairment. The decrease in PAK in the brain of Alzheimer's disease (AD) patients is suspected to underlie synaptic and dendritic disturbances associated with its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service