ABE446
Anti-Monomethyl Histone H1 (Lys26) Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
Histone H1.3, Histone H1c, Histone H1s-2
About This Item
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
target post-translational modification
monomethylation (Lys26)
Gene Information
human ... HIST1H1D(3007)
General description
Specificity
Immunogen
Application
Dot Blot Specificity Analysis: This antibody detected Monomethyl Histone H1 (Lys26) in in an Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and in an Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).
Quality
Western Blotting Analysis: 0.25 µg/mL of this antibody detected Monomethyl Histone H1 (Lys26) in 10 µg of HeLa acid extract.
Target description
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service