Skip to Content
Merck
All Photos(1)

Key Documents

AB9287

Sigma-Aldrich

Anti-Somatostatin Receptor Type 5 Antibody

Chemicon®, from rabbit

Synonym(s):

SSTR5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SSTR3(6753)

Specificity

Recognizes Somatostatin Receptor Type 5 [SSTR5].

Immunogen

Synthetic peptide from the C-terminal domain of human Somatostatin Receptor Type 5.

Application

Anti-Somatostatin Receptor Type 5 Antibody detects level of Somatostatin Receptor Type 5 & has been published & validated for use in IH(P).

Physical form

Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.1% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofie Eline Tollefsen et al.
Cancers, 13(22) (2021-11-28)
Meningiomas have high recurrence rates despite frequently benign histopathological appearances. Somatostatin receptors (SSTRs) may be reliable biomarkers that could identify patients with increased risk of recurrence. Even though SSTRs are previously detected in meningiomas, their associations to clinicopathological features remain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service