Skip to Content
Merck
All Photos(1)

Documents

AB5871

Sigma-Aldrich

Anti-SNAP-25 Antibody

Chemicon®, from goat

Synonym(s):

Synaptosomal-associated Protein 25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SNAP25(6616)

Specificity

SNAP-25

Immunogen

Recombinant human SNAP-25

Application

Anti-SNAP-25 Antibody is an antibody against SNAP-25 for use in IP, WB, IH.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blot: 1:100 - 1:2000
ELISA: 1:1000 - 1:100,000
Immunohistochemistry: 1:50 - 1:100

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
IgG fraction (protein A) in PBS with 50% Glycerol and 0.01% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months from date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoit J Gentil et al.
Journal of neurochemistry, 131(5), 588-601 (2014-08-13)
Peripherin is a type III intermediate filament protein, the expression of which is associated with the acquisition and maintenance of a terminally differentiated neuronal phenotype. Peripherin up-regulation occurs during acute neuronal injury and in degenerating motor neurons of amyotrophic lateral
Arek Szklarczyk et al.
Journal of neurochemistry, 102(4), 1256-1263 (2007-05-03)
Matrix metalloproteinases (MMPs) belong to a family of zinc dependent enzymes best studied for their role in cancer and inflammation. Though MMPs typically target extracellular proteins, here we show that MMP-7, an MMP family member which lacks a C-terminal hemopexin-like
Kwok-Ho Lam et al.
Cell reports, 30(8), 2526-2539 (2020-02-27)
Botulinum neurotoxin (BoNT) is one of the most acutely lethal toxins known to humans, and effective treatment for BoNT intoxication is urgently needed. Single-domain antibodies (VHH) have been examined as a countermeasure for BoNT because of their high stability and
Noelle C Anastasio et al.
Journal of neurochemistry, 113(6), 1504-1515 (2010-03-30)
The action of serotonin (5-HT) at the 5-HT(2C) receptor (5-HT(2C)R) in cerebral cortex is emerging as a candidate modulator of neural processes that mediate core phenotypic facets of several psychiatric and neurological disorders. However, our understanding of the neurobiology of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service