Skip to Content
Merck
All Photos(1)

Key Documents

AB5727

Sigma-Aldrich

Anti-Sox10 Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-Sox10 antibody, Sox10 marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Sox 10 is part of a family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. This protein acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Sox 10 is expressed in multipotent neural crest stem cells, and its expression is downregulated upon their differentiation. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.

Specificity

Sox10. The immunogen sequence is 70% conserved with Sox8 and Sox9 and therefore the antibody may show cross reactivity to these proteins.

Immunogen

Synthetic peptide from Sox10.

Application

Anti-Sox10 Antibody detects level of Sox10 & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Neuronal & Glial Markers
Western blot: 0.1-0.5 μg/mL using ECL on fetal mouse brain tissue lysate. The antibody reacts with proteins of ~60 kDa. An additional band of ~120 kDa may also be seen depending on sample used.

Optimal working dilutions must be determined by the end user.

Target description

Approx. 55 kDa

Linkage

Replaces: AB5774

Physical form

Affinity purified immunoglobulin. Liquid.

Storage and Stability

Maintain at +2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human embryonic stem cell-derived oligodendrocyte progenitor cell transplants remyelinate and restore locomotion after spinal cord injury.
Keirstead, Hans S, et al.
The Journal of Neuroscience, 25, 4694-4705 (2005)
Débora Guerini Souza et al.
Journal of cellular biochemistry, 118(10), 3111-3118 (2017-04-05)
Astrocytes are versatile cells involved in synaptic information processing, energy metabolism, redox homeostasis, inflammatory response, and structural support of the brain. Recently, we established a routine protocol of cultured astrocytes derived from adult and aged Wistar rats, which present several
Kyohei Fujita et al.
The FEBS journal, 283(20), 3791-3806 (2016-10-21)
Neural crest-derived stem cells (NCSCs) are tissue-specific stem cells derived from multipotent neural crest cells. NCSCs are present in some adult tissues such as dorsal root ganglia, sciatic nerve, and bone marrow. However, little is known about the formation mechanisms
Jessica Jacobs-Li et al.
eLife, 12 (2023-10-25)
During development, much of the enteric nervous system (ENS) arises from the vagal neural crest that emerges from the caudal hindbrain and colonizes the entire gastrointestinal tract. However, a second ENS contribution comes from the sacral neural crest that arises
Na Wang et al.
Glia, 70(11), 2093-2107 (2022-07-02)
In humans, loss-of-function mutations of Kcnj10 in SeSAME/EAST syndrome, which encodes the inwardly rectifying K+ channel 4.1 (Kir 4.1), causes progressive neurological decline. Despite its rich expression in oligodendrocyte (OL) lineage cells and an emerging link with demyelinating disease, the

Articles

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Derivation and characterization of functional human neural stem cell derived oligodendrocyte progenitor cells (OPCs) that efficiently myelinate primary neurons in culture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service