Skip to Content
Merck
All Photos(1)

Key Documents

AB1779SP

Sigma-Aldrich

Anti-Brain Derived Neurotrophic Factor Antibody

Chemicon®, from rabbit

Synonym(s):

BDNF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

Specificity

Brain Derived Neurotrophic Factor (BDNF). By dot blot, less than 1% cross-reactivity against NGF, NT3 or NT4.

Immunogen

Recombinant Human Brain Derived Neurotrophic Factor (BDNF).

Application

Anti-Brain Derived Neurotrophic Factor Antibody detects level of Brain Derived Neurotrophic Factor & has been published & validated for use in ELISA, WB, IH.
Immunohistochemistry: 5-10μg/mL (see suggested protocol).

Immunoblotting: 5-10μg/mL. BDNF is not easily extracted in standard buffers. Dissected tissues should be homogenized in 10 volumes of 100mM phosphate buffer containing 1mM EDTA, 2M guanidine hydrochloride (pH7.2), and three protease inhibitors, 10mM N-ethylmaleimide, 0.36mM pepstatin, and 1mM PMSF. Homogenates are then sonicated and centrifuged at 46K x g for 30 minutes at 4C. BDNF migrates as 18kDa, with a homodimer at ~27-30kDa. 1:1,000

ELISA: 5-10μg/mL

BIOLOGICAL ACTIVITY: Neutralizes BDNF, but not other neurotrophins.



Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Neuroinflammation & Pain

Linkage

Replaces: AB1534P

Physical form

Format: Purified
Immunoglobulin fraction lyophilized from PBS, with no preservatives. Reconstitute with 500 μL of sterile distilled water (1 mg/mL).

Storage and Stability

Maintain lyophilized material at -20 to -70°C for up to one year after date of receipt. After reconstitution maintain at -20 to -70°C in undiluted aliquots for up to six months. Glycerol (ACS grade or better) can be added (1:1) for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Nikulina et al.
Neuroscience, 212, 38-48 (2012-04-24)
Intermittent social defeat stress exposure augments behavioral response to psychostimulants in a process termed cross-sensitization. Brain-derived neurotrophic factor (BDNF) mediates synaptic plasticity and cellular responses to stress and drugs of abuse. We previously showed that repeated social defeat stress persistently
BDNF overexpression in the ventral tegmental area prolongs social defeat stress-induced cross-sensitization to amphetamine and increases ?FosB expression in mesocorticolimbic regions of rats.
Wang, J; Fanous, S; Terwilliger, EF; Bass, CE; Hammer, RP; Nikulina, EM
Neuropsychopharmacology null
Nicolás Unsain et al.
Journal of neurochemistry, 111(2), 428-440 (2009-08-19)
Brain-derived neurotrophic factor (BDNF) is involved in many aspects of neuronal biology and hippocampal physiology. Status epilepticus (SE) is a condition in which prolonged seizures lead to neuronal degeneration. SE-induced in rodents serves as a model of Temporal Lobe Epilepsy
Junshi Wang et al.
Neuropharmacology, 109, 121-130 (2016-05-08)
Stress is a major risk factor for substance abuse. Intermittent social defeat stress increases drug self-administration (SA) and elevates brain-derived neurotrophic factor (BDNF) expression in the ventral tegmental area (VTA) in rats. Intra-VTA BDNF overexpression enhances social defeat stress-induced cross-sensitization
Alfonso Paredes et al.
Developmental biology, 267(2), 430-449 (2004-03-12)
Although it is well established that both follicular assembly and the initiation of follicle growth in the mammalian ovary occur independently of pituitary hormone support, the factors controlling these processes remain poorly understood. We now report that neurotrophins (NTs) signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service