Skip to Content
Merck
All Photos(1)

Key Documents

AB15242

Sigma-Aldrich

Anti-zDHHC8 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Zinc finger DHHC domain-containing protein 8, Zinc finger protein 378, membrane-associated DHHC8 zinc finger protein, zinc finger, DHHC domain containing 8, zinc finger, DHHC domain like containing 1, zinc finger, DHHC-type containing 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

species reactivity (predicted by homology)

horse, bovine, Xenopus, chicken, pig, chimpanzee, ape, opossum, mouse, dog

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

chimpanzee ... Zdhhc8(458662)
human ... ZDHHC8(29801)

General description

Six genes in the 1.5 Mb region of chromosome 22 deleted in DiGeorge/22q11 deletion syndrome–Mrpl40, Prodh, Slc25a1, Txnrd2, T10, and Zdhhc8–encode mitochondrial proteins. All six genes are expressed in the
brain, and maximal expression coincides with peak forebrain synaptogenesis shortly after birth. Furthermore, their protein products are associated with brain mitochondria, including those in synaptic terminals. Among the six, only Zddhc8 influences mitochondria-regulated apoptosis when overexpressed, and appears to interact biochemically with established mitochondrial proteins. Zdhhc8 has an apparent interaction with Uqcrc1, a component of mitochondrial complex III. The two proteins are coincidently expressed in pre-synaptic processes; however, Zdhhc8 is more frequently seen in glutamatergic terminals (1).

Specificity

Cat. AB15242 recognizes the C-terminus of zDHHC8.

Immunogen

Epitope: C-Terminus
Syntheic linear peptide.

Application

Anti-zDHHC8 Antibody is an antibody against zDHHC8 for use in WB.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Routinely evaluated by western blot in fetal human brain lysate.

Target description

~71 kDa

Physical form

ImmunoAffinity Purified
Purified in PBS with 0.05% NaN3

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Handling Recommendations: Prior to removing the cap, gently centrifuge the vial and gently mix the solution.

Analysis Note

Control
Fetal human brain

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service