Skip to Content
Merck
All Photos(1)

Key Documents

8.41470

Sigma-Aldrich

Creatine monohydrate

for synthesis

Synonym(s):

Creatine monohydrate, N-Amidinosarcosine monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9N3O2 · xH2O
CAS Number:
Molecular Weight:
131.13 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-306-6
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

form

powder

autoignition temp.

>800 °C

pH

6.9 (20 °C, 10 g/L in H2O)

mp

296-297 °C (decomposition)

solubility

17 g/L

storage temp.

2-30°C

SMILES string

N(CC(=O)O)(C)\C(=N\[H])\N.O

InChI

1S/C4H9N3O2.H2O/c1-7(4(5)6)2-3(8)9;/h2H2,1H3,(H3,5,6)(H,8,9);1H2

InChI key

MEJYXFHCRXAUIL-UHFFFAOYSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service