Skip to Content
Merck
All Photos(1)

Key Documents

8.18150

Sigma-Aldrich

Tin(II) chloride

anhydrous for synthesis

Synonym(s):

Tin(II) chloride, Tin dichloride, Stannous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Sn
CAS Number:
Molecular Weight:
189.62
UNSPSC Code:
12352302
EC Index Number:
231-868-0
NACRES:
NA.22

Quality Level

form

powder

potency

700 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: reductant

pH

2.0 (20 °C, 100 g/L in H2O)

bp

623 °C/1013 hPa

mp

246 °C

solubility

2700 g/L

density

3.95 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/2ClH.Sn/h2*1H;/q;;+2/p-2

InChI key

AXZWODMDQAVCJE-UHFFFAOYSA-L

Analysis Note

Assay (iodometric): ≥ 97.0 %
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service