Skip to Content
Merck
All Photos(2)

Key Documents

8.14181

Sigma-Aldrich

Bis(2-ethylhexyl) phosphate

for synthesis

Synonym(s):

Bis(2-ethylhexyl) phosphate, HDEHP, Phosphoric acid bis (2-ethylhexyl) ester, Bis(2-ethylhexyl) phosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3(CH2)3CH(C2H5)CH2O)2POOH
CAS Number:
Molecular Weight:
322.42
MDL number:
UNSPSC Code:
12352106
EC Index Number:
206-056-4
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (acidimetric)

autoignition temp.

255 °C

potency

4940 mg/kg LD50, oral (Rat)
1250 mg/kg LD50, skin (Rabbit)

refractive index

n25/589 1.4420

pH

3 (1 g/L in H2O)

bp

48 °C/16 hPa

mp

-50 °C

transition temp

flash point 150 °C

solubility

<1 g/L

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[P](=O)(OCC(CCCC)CC)(OCC(CCCC)CC)O

InChI key

SEGLCEQVOFDUPX-UHFFFAOYSA-N

Application

  • Production of highly purified zirconium oxide from zircon mineral leach liquor using bis-(2-ethylhexyl) phosphate as a promising extractant: This research evaluates the feasibility of using bis-(2-ethylhexyl) phosphate for zirconium separation, demonstrating its potential in zirconium oxide production (AH Ali, 2022).
  • Optimization of indium recovery and separation from LCD waste by solvent extraction with bis (2-ethylhexyl) phosphate (D2EHPA): This study explores solvent extraction methods for indium recovery from LCD waste, highlighting the efficiency of bis(2-ethylhexyl) phosphate (J Yang, C Ekberg, T Retegan, 2014).
  • Homogeneous liquid–liquid extraction of metal ions with non-fluorinated bis (2-ethylhexyl) phosphate ionic liquids having a lower critical solution temperature: This article investigates the use of bis(2-ethylhexyl) phosphate ionic liquids for metal ion extraction, contributing to the development of more environmentally friendly extraction technologies (D Depuydt, L Liu, C Glorieux, W Dehaen, 2015).

Analysis Note

Assay (acidimetric): ≥ 95.0 %
Density (d 20 °C/ 4 °C): 0.968 - 0.975
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

266.0 °F

Flash Point(C)

130 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service