Skip to Content
Merck
All Photos(1)

Key Documents

8.01555

Sigma-Aldrich

Butyraldehyde

for synthesis

Synonym(s):

Butyraldehyde, Butanal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
MDL number:
UNSPSC Code:
12352114
EC Index Number:
204-646-6
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

116 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

190 °C

potency

2490 mg/kg LD50, oral (Rat)
2848 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-11.1 % (v/v)

pH

6-7 (20 °C, 71 g/L in H2O)

bp

75-76 °C/1013 hPa

mp

-99 °C

transition temp

flash point -12 °C

density

0.80 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=CCCC

InChI

1S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3

InChI key

ZTQSAGDEMFDKMZ-UHFFFAOYSA-N

Application

  • Advanced tetra amino (ATA-100) cobalt(II) phthalocyanine-based metallo-covalent organic polymer: Utilizing butyraldehyde in the synthesis of cobalt(II) phthalocyanine-based polymers, this research provides an innovative approach for detecting volatile organic compounds, presenting significant implications for environmental monitoring and safety assessments in various industries (Güney et al., 2023).
  • Catalytic production of bio-renewable butyl butyrate: This review emphasizes the catalytic use of butyraldehyde for producing butyl butyrate, a potential jet fuel blend, highlighting its role in sustainable fuel development and its importance in the biofuel industry (Kushwaha et al., 2022).
  • Detection of aliphatic aldehydes: A pillar[5]arene-based fluorescent supramolecular polymer demonstrates vaporchromic behavior, which is essential for detecting butyraldehyde among other aldehydes. This technology is crucial for enhancing safety protocols in chemical manufacturing environments (Shi et al., 2022).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.802 - 0.804
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

50.0 °F - Pensky-Martens closed cup

Flash Point(C)

< 10 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service