Skip to Content
Merck
All Photos(1)

Key Documents

8.00259

Sigma-Aldrich

Allyl 2,3-epoxypropyl ether

for synthesis

Synonym(s):

Allyl 2,3-epoxypropyl ether, Allyl glycidyl ether, Glycidyl allyl ether, 1-(Allyloxy)-2,3-epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-442-4
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

2.6 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

264 °C

potency

922 mg/kg LD50, oral (Rat)
2550 mg/kg LD50, skin (Rabbit)

bp

154 °C/1013 hPa

mp

-100 °C

transition temp

flash point 45 °C

density

0.97 g/cm3 at 20 °C

SMILES string

O1C(C1)COCC=C

InChI

1S/C6H10O2/c1-2-3-7-4-6-5-8-6/h2,6H,1,3-5H2

InChI key

LSWYGACWGAICNM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.967 - 0.969
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service