Skip to Content
Merck
All Photos(1)

Key Documents

693023

Millipore

Zwittergent® 3-16 Detergent

A synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a broad pH range.

Synonym(s):

n-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H45NO3S
CAS Number:
Molecular Weight:
391.65
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

Quality Level

Assay

≥99% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

aggregation number

155

color

white

CMC

10 - 60 μM

solubility

water: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])CCC[N+](CCCCCCCCCCCCCCCC)(C)C

InChI

1S/C21H45NO3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22(2,3)20-18-21-26(23,24)25/h4-21H2,1-3H3

InChI key

TUBRCQBRKJXJEA-UHFFFAOYSA-N

General description

A synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a broad pH range. This property is attributed to the presence of a strongly basic quaternary ammonium ion and an acidic sulfonate ion of equal strength. This ionic balance presumably prevents irreversible binding to either anionic or cationic compounds.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (4°C). Stock solutions are stable for up to 1 month at 4°C.

Other Notes

Sekine, M., et al. 1990. J. Biochem. 108, 103.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Zwittergent is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service