Skip to Content
Merck
All Photos(1)

Documents

616464

Sigma-Aldrich

SB431542

≥97% (HPLC), liquid, TGF-β RI kinase inhibitor, Calbiochem®

Synonym(s):

InSolution TGF-β RI Kinase Inhibitor VI, SB431542

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N4O3 · 2H2O
CAS Number:
Molecular Weight:
420.42
UNSPSC Code:
51111800
NACRES:
NA.77

product name

TGF-β RI Kinase Inhibitor VI, SB431542, InSolution, ≥97%

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−20°C

InChI

1S/C22H16N4O3/c23-21(27)13-4-6-14(7-5-13)22-25-19(20(26-22)16-3-1-2-10-24-16)15-8-9-17-18(11-15)29-12-28-17/h1-11H,12H2,(H2,23,27)(H,25,26)

InChI key

FHYUGAJXYORMHI-UHFFFAOYSA-N

General description

A cell-permeable triarylimidazole compound that is shown to effectively inhibit cellular Smad2 phosphorylation (>90% inhibition by 10 µM inhibitor) upon vector-mediated expression of constitutively active ALK4, ALK5, or ALK7 in NIH 3T3 cells, while exhibiting little effect against Smad1 phosphorylation by other members of type I receptors for TGF-β in NIH 3T3 cultures expressing active ALK1, 2, 3, or 6. When tested directly in cell-free kinase assays, SB431542 is demonstrated to potently inhibit the activity of ALK4 and ALK5 (IC50 = 140 nM and 94 nM, respectively) with no or much reduced potency toward a panel of 24 other kinases (IC50 ≥10 µM in the presence of 10 µM ATP), including ALK2 and ALK6. Reported to improve the efficiency of 4-TF-induced human iPSCs generation from fibroblast cultures by >200-fold when used together with PD0325901 (Cat. No. 444966) and Thiazovivin (Cat. No. 420220).

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 100 mM (5 mg/119 µl) solution of SB431542 (Cat. No. 616461) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Ikushima, H., et al. 2009. Cell Stem Cell5, 504.
Lin, T., et al. 2009. Nat. Methods6, 805.
Maherali, N. and Hochedlinger, K., 2009. Curr. Biol.19, 1718.
Callahan, J.F., et al. 2002. J. Med. Chem.45, 999.
Inman, G.J., et al. 2002. Mol. Pharmacol.62, 65.
Laping, N.J., et al. 2002. Mol. Pharmacol.62, 58.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mine Koprulu et al.
The Journal of clinical endocrinology and metabolism, 107(4), 1065-1077 (2021-12-08)
Biological and translational insights from large-scale, array-based genetic studies of fat distribution, a key determinant of metabolic health, have been limited by the difficulty in linking predominantly noncoding variants to specific gene targets. Rare coding variant analyses provide greater confidence
Jorian D Hapeman et al.
BMC ecology and evolution, 23(1), 39-39 (2023-08-22)
In spite of extensive research, cancer remains a major health problem worldwide. As cancer progresses, cells acquire traits that allow them to disperse and disseminate to distant locations in the body - a process known as metastasis. While in the
Ming-An Sun et al.
Molecular biology and evolution, 38(11), 4992-5004 (2021-07-29)
In mammals, the placenta mediates maternal-fetal nutrient and waste exchange and acts in an immunomodulatory way to facilitate maternal-fetal tolerance. The placenta is highly diverse across mammalian species, yet the molecular mechanisms that distinguish the placenta of human from other
Xin-Yi Xu et al.
Frontiers in molecular biosciences, 9, 835508-835508 (2022-03-05)
Hepatic stellate cells (HSCs) play an essential role in the development of liver fibrosis. Antrodia camphorata (A. camphorata) is a medicinal fungus with hepatoprotective effect. This study investigated whether Antrodin C, an A. camphorata-fermented metabolite, could exert a protective role
Chao-Min Cheng et al.
The Journal of pathology, 240(1), 38-49 (2016-05-29)
MicroRNAs (miRNAs) are involved in the tumourigenesis of various cancers by regulating their downstream targets. To identify the changes of miRNAs in oral squamous cell carcinoma (OSCC), we investigated the expression profiles of miRNAs in 40 pairs of OSCC specimens

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service