Skip to Content
Merck
All Photos(1)

Key Documents

539128

Sigma-Aldrich

Protease Inhibitor Kit

solid, for the inhibition of serine, cysteine, trypsin-like, aspartic and chymotrypsin proteases. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
form:
solid
solubility:
water: soluble
storage temp.:
−20°C
Pricing and availability is not currently available.

Product Name

Protease Inhibitor Set, The Protease Inhibitor Set controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Contains 50 mg of AEBSF, HCl (Cat. No. 101500), 1 mg of E-64 (Cat. No. 324890), 1 mg of EST (E-64d; Cat. No. 330005), 5 mg of Leupeptin, Hemisulfate (Cat. No. 108975), 5 mg of Pepstatin A, Synthetic (Cat. No. 516481), 50 mg of TLCK, HCl (Cat. No. 616382) and 250 mg of TPCK (Cat. No. 616387).

Specificity

Inhibits protease activity when the precise nature of proteolytic activity is not known.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Other Notes

Munger, J.S., et al. 1995. Biochem. J.311, 299.
Murao, S., et al. 1993. J. Biol. Chem.268, 349.
Schmidt, H.H., et al. 1992. Mol. Pharmacol.41, 615
Szewczuk, Z., et al. 1992. Int. J. Peptide Res.40, 233.
McGowan, E.B., et al. 1989. Biochem. Biophys. Res. Commun.158, 432.
Brass, L.F., and Shaltil, S.J. 1988. J. Biol. Chem.263, 5210.
Barrett, A.J., et al. 1982. Biochem. J.201, 189.
Hanada, K. 1978. Agric. Biol. Chem.42, 523.
Umezawa, H., 1976. Methods Enzymol.45, 678.
Markwardt, F., et al. 1973. Thromb. Res.2, 343.
Shaw, E. 1967. Methods Enzymol.11, 677.
Shaw, E., et al. 1965. Biochemistry4, 2219.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service