Skip to Content
Merck
All Photos(1)

Key Documents

5.08313

Sigma-Aldrich

HSP90α Inhibitor, PU-29F

Synonym(s):

HSP90α Inhibitor, PU-29F, 2-Fluoro-9-(2-isopropoxyethyl)-8-(3,4,5-trimethoxybenzyl)-9H-purin-6-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26FN5O4
CAS Number:
Molecular Weight:
419.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable 6-amino-purine based compound that selectively binds to HSP90a (EC50 = 5.4 µM) and inhibits its activity in SKBr3 and MCF-7 breast cancer cells. Exhibits much reduced activity towards other paralogs (EC50 = 22.85, 71, and 173 5.4 µM for HSP90b, GRP94 AND Trap-1, respectively). Selectively depletes cytosolic HER2 and induces its redistribution towards lysosomes and early endosomes (~20 5.4 µM) without affecting membrane associated HER2. Does not appear to be toxic towards nonmalignant cells.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HSP90a

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Patel, P.D., et al. 2013. Nat. Chem. Biol.9, 677.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service