5.06125
Paroxetine Hydrochloride
Synonym(s):
Paroxetine Hydrochloride, 5-HT transporter, Paroxetine, Paxil
About This Item
Recommended Products
Assay
≥99% (HPLC)
Quality Level
form
solid
potency
0.065 nM Ki
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: soluble
storage temp.
2-8°C
SMILES string
Fc1ccc(cc1)[C@H]2[C@@H](CNCC2)COc3cc4c(cc3)OCO4
InChI
1S/C19H20FNO3/c20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18/h1-6,9,14,17,21H,7-8,10-12H2/t14-,17-/m0/s1
InChI key
AHOUBRCZNHFOSL-YOEHRIQHSA-N
Related Categories
General description
Biochem/physiol Actions
5-HT Uptake Transporter
Warning
Reconstitution
Other Notes
Bourin, M., et al. 2001. CNS Drug Rev.7, 25.
Thomas, R., et al. 1997. Psychopharmacology (Berl).93, 193.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service