Skip to Content
Merck
All Photos(1)

Key Documents

487655

Sigma-Aldrich

p-Nitrophenyl Phosphate, DiTris Salt

Excellent substrate for alkaline phosphatase-based ELISA assays.

Synonym(s):

p-Nitrophenyl Phosphate, DiTris Salt, pNPP, 2Tris

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28N3O12P
CAS Number:
Molecular Weight:
461.36
MDL number:
UNSPSC Code:
41116158
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.05% Free-p-Nitrophenol

color

off-white to pale yellow

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

[P](=O)([O-])([O-])Oc1ccc(cc1)[N+](=O)[O-].[N+H3]C(CO)(CO)CO.[N+H3]C(CO)(CO)CO

InChI

1S/C6H6NO6P.2C4H11NO3/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;2*5-4(1-6,2-7)3-8/h1-4H,(H2,10,11,12);2*6-8H,1-3,5H2

InChI key

XXAXKCWOTRABOW-UHFFFAOYSA-N

General description

Excellent substrate for alkaline phosphatase-based ELISA assays. Produces a yellow soluble end product that can be read at 405 nm to 410 nm. Both kinetic and end point determinations can be performed.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate for alkaline phosphatase-based ELISA assays
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service