Skip to Content
Merck
All Photos(1)

Key Documents

444291

Sigma-Aldrich

MMP408

≥94% (HPLC), solid, MMP-12 inhibitor, Calbiochem®

Synonym(s):

MMP-12 Inhibitor, MMP408, (S)-2-(8-(Methoxycarbonylamino)dibenzo[b,d]furan-3-sulfonamido)-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O7S
CAS Number:
Molecular Weight:
420.44
UNSPSC Code:
12352200
NACRES:
NA.77

product name

MMP-12 Inhibitor, MMP408, The MMP-12 Inhibitor, MMP408, also referenced under CAS 1258003-93-8, controls the biological activity of MMP-12. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Quality Level

Assay

≥94% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A dibenzofuranylsulfanamido-valine compound that acts as a potent, active site zinc-targeting, MMP-12-selective inhibitor (IC50 = 2 nM against human MMP-12) with much weaker activity against MMP-12 from mouse/rat/sheep species (IC50 = 160, 320 and 22.3 nM , respectively) or human MMP-3/13/14 (IC50 = 351, 120 and 1100 nM , respectively), and practically ineffective toward human Agg-1/2 (IC50 >5 µM), TACE (IC50 >25 µM), and MMP-1/7 (IC50 >6 µM). MMP408 is orally active in mice and shown to effectively alleviate rhMMP-12-induced pulmonary inflammatory response in mice in vivo by more than 50% (5 mg/kg, p.o., b.i.d.).
A dibenzofuranylsulfanamido-valine compound that acts as a potent, active site zinc-targeting, MMP-12-selective inhibitor (IC50 = 2 nM against human MMP-12) with much weaker activity against MMP-12 from mouse/rat/sheep species (IC50 = 160, 320 and 22.3 nM, respectively) or human MMP-3/13/14 (IC50 = 351, 120 and 1100 nM , respectively), and practically ineffective toward human Agg-1/2 (IC50 >5 µM), TACE (IC50 >25 µM), and MMP-1/7 (IC50 >6 µM). MMP408 is orally active in mice and shown to effectively alleviate rhMMP-12-induced pulmonary inflammatory response in mice in vivo by more than 50% (5 mg/kg, p.o., b.i.d.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Li, W., et al. 2009. J. Med. Chem.52, 1799.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caijiao Yi et al.
Journal of neuroinflammation, 19(1), 78-78 (2022-04-07)
Macular subretinal fibrosis is the end-stage complication of neovascular age-related macular degeneration (nAMD). We previously developed a mouse model of two-stage laser-induced subretinal fibrosis that mimics closely the dynamic course of macular fibrosis in nAMD patients. This study was aimed
Lingbi Jiang et al.
BMC cancer, 21(1), 1297-1297 (2021-12-06)
Colorectal cancer is a malignant gastrointestinal cancer, in which some advanced patients would develop cancer cachexia (CAC). CAC is defined as a multi-factorial syndrome characterized by weight loss and muscle loss (with or without fat mass), leading to progressive dysfunction
Meghali Nighot et al.
Journal of Crohn's & colitis, 15(10), 1751-1765 (2021-04-10)
Matrix metalloproteinases [MMPs] play an important role in extracellular matrix regulation during cell growth and wound healing. Increased expression of MMP-12 [human macrophage elastase] has been reported in inflammatory bowel disease [IBD] which is characterised by the loss of epithelial
Airi Makino et al.
iScience, 24(10), 103201-103201 (2021-10-28)
Respiratory syncytial virus (RSV) infection often exacerbates bronchial asthma, but there is no licensed RSV vaccine or specific treatments. Here we show that RSV-induced alveolar macrophages, which produce high levels of matrix metalloproteinase-12 (MMP-12), exacerbate allergic airway inflammation with increased
Hao Huang et al.
Cell reports, 36(2), 109363-109363 (2021-07-15)
Although activating mutations of the anaplastic lymphoma kinase (ALK) membrane receptor occur in ∼10% of neuroblastoma (NB) tumors, the role of the wild-type (WT) receptor, which is aberrantly expressed in most non-mutated cases, is unclear. Both WT and mutant proteins

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service