Skip to Content
Merck
All Photos(1)

Key Documents

401393

Sigma-Aldrich

Goat Anti-Rabbit IgG, H & L Chain Specific Peroxidase Conjugate

liquid, Calbiochem®

Synonym(s):

Goat IgG antibody, Goat anti-rabbit IgG, Peroxidase conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

0.02% thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Adsorbed against bovine, horse, human, and mouse serum proteins.
Immunoaffinity purified goat polyclonal antibody adsorbed against bovine, horse, human, and mouse serum proteins and conjugated to horseradish peroxidase. Recognizes rabbit IgG, heavy and light chains.
This Goat Anti-Rabbit IgG, H & L Chain Specific Peroxidase Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Rabbit IgG, H & L Chain Specific.

Immunogen

Rabbit

Application

Enzyme Immunoassay (1:10,000)

Immunoelectrophoresis (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.

Other Notes

Monospecific for rabbit IgG, heavy and light chains, as determined by immunoelectrophoresis against normal rabbit serum. Cross-reactivity with normal bovine, horse, human, and mouse serum: <2% by direct solid phase immunoassay. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niku K J Oksala et al.
Redox biology, 3, 25-28 (2014-12-03)
Heat shock proteins (HSPs), originally identified as heat-inducible gene products, are a family of highly conserved proteins that respond to a wide variety of stress including oxidative stress. Although both acute and chronic oxidative stress have been well demonstrated to
Yotam Blech-Hermoni et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 245(8), 854-873 (2016-05-05)
CUG-BP, Elav-like family member 1 (CELF1) is a multifunctional RNA binding protein found in a variety of adult and embryonic tissues. In the heart, CELF1 is found exclusively in the myocardium. However, the roles of CELF1 during cardiac development have
Carla L Busceti et al.
Frontiers in pharmacology, 8, 111-111 (2017-03-30)
Dkk-3 is a member of the dickkopf protein family of secreted inhibitors of the Wnt pathway, which has been shown to enhance angiogenesis. The mechanism underlying this effect is currently unknown. Here, we used cultured HUVECs to study the involvement
Chunsheng Liu et al.
Circulation research, 97(2), 135-143 (2005-06-25)
Recent experimental studies suggest C-reactive protein (CRP) may be a potential mediator of atherosclerosis and its complications. However, there is growing criticism of in vitro CRP studies that use commercial CRP preparations containing biologically active contaminants. The effects of commercial
Umberto Rodella et al.
Disease models & mechanisms, 10(5), 597-603 (2017-01-10)
The neuromuscular junction is exposed to different types of insult, including mechanical trauma, toxins and autoimmune antibodies and, accordingly, has retained through evolution a remarkable ability to regenerate. Regeneration is driven by multiple signals that are exchanged among the cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service