Skip to Content
Merck
All Photos(1)

Key Documents

341310

Sigma-Aldrich

FABP4 Inhibitor

The FABP4 Inhibitor, also referenced under CAS 300657-03-8, controls the biological activity of FABP4.

Synonym(s):

FABP4 Inhibitor, adipocyte Fatty-Acid-Binding Protein Inhibitor, ((2ʹ-(5-Ethyl-3,4-diphenyl-1H-pyrazol-1-yl)(1,1ʹ-biphenyl)-3-yl)oxy)-acetic acid, BMS309403, BMS-309403, BMS 309403

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H26N2O3
CAS Number:
Molecular Weight:
474.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 20 mg/mL
DMSO: 50 mg/mL
methanol: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]3(nc(c(c3CC)c5ccccc5)c4ccccc4)c1c(cccc1)c2cc(ccc2)OCC(=O)O

InChI

1S/C31H26N2O3/c1-2-27-30(22-12-5-3-6-13-22)31(23-14-7-4-8-15-23)32-33(27)28-19-10-9-18-26(28)24-16-11-17-25(20-24)36-21-29(34)35/h3-20H,2,21H2,1H3,(H,34,35)

InChI key

SJRVJRYZAQYCEE-UHFFFAOYSA-N

General description

A cell-permeable biphenylazolo-oxyacetate that acts as a potent and selective inhibitor of adipocyte Fatty-Acid-Binding Protein (aFABP/aP2) by targeting its fatty acid-binding pocket (Ki = <2 nM in a competitive binding assay using 1,8-ANS), while exhibiting much lower affinity for muscle and epidermal FABP′s (Ki = 250 nM and 350 nM, respectively). Shown to effectively block foam cell transformation and the cellular expression of inflammatory mediators in aP2-positive, but not aP2-negative, THP-1 macrophage in vitro, as well as improve the glucose metabolism and insulin sensitivity in mice in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Furuhashi, M., et al. 2007. Nature447, 959.
Sulsky, R., et al. 2007. Bioorg. Med. Chem. Lett.17, 3511.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunyan Yu et al.
Cancer immunology, immunotherapy : CII, 69(1), 115-126 (2019-12-06)
Pro-inflammatory cytokines are crucial mediators of cancer development, representing potential targets for cancer therapy. The molecular mechanism of a vital pro-inflammatory cytokine, IL-17A, in cancer progression and its potential use in therapy through influencing fatty acid (FA) metabolism, especially FA
Sanjay Basak et al.
Cell biology international, 44(5), 1237-1251 (2020-02-20)
Curcumin has a protective role in placental diseases like preeclampsia and preterm birth. Very little is known about its functional effects on growth, angiogenesis, and epigenetic activities of human first trimester placenta. HTR8/SVneo trophoblasts cells were used as model for
Qiuyuan Zhu et al.
Cell biology international, 39(5), 540-547 (2015-01-13)
The aims of this study were to delineate the expression of fatty-acid binding protein (FABP) 4 in human uterine endometrium and its function in the regulation of proliferation, migration and invasion of epithelial cells. Immunohistochenistry, immunofluorence and Western blotting were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service