Skip to Content
Merck
All Photos(1)

Documents

252718

Millipore

Decyl-β-D-maltopyranoside

ULTROL® Grade, ≥99% (HPLC), non-ionic detergent for solubilization of membrane-bound proteins

Synonym(s):

n-Decyl-β-D-maltopyranoside, ULTROL® Grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21

Quality Level

Assay

≥99% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

CMC

1.6 mM

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C22H42O11/c1-2-3-4-5-6-7-8-9-10-30-21-19(29)17(27)20(14(12-24)32-21)33-22-18(28)16(26)15(25)13(11-23)31-22/h13-29H,2-12H2,1H3/t13-,14-,15-,16+,17-,18-,19-,20-,21-,22-/m1/s1

InChI key

WOQQAWHSKSSAGF-WXFJLFHKSA-N

General description

Absorbance (1%, H2O, 260 nm): <0.1.
Non-ionic detergent designed for solubilization of membrane-bound proteins.
Non-ionic detergent designed for the solubilization of membrane-bound proteins (absorbance: <0.1 at 260 nm, 1% H2O; CMC: 1.6 mM).

Application


  • A chromatographic network for the purification of detergent-solubilized six-transmembrane epithelial antigen of the prostate 1 from Komagataella pastoris mini-bioreactor lysates.: This research highlights the effective use of Decyl-β-D-maltopyranoside in the purification processes for proteins extracted from yeast, improving yield and purity essential for biochemical research and pharmaceutical applications (Barroca-Ferreira et al., 2022).

  • The Caco-2 cell culture model enables sensitive detection of enhanced protein permeability in the presence of N-decyl-β-d-maltopyranoside.: Exploring the effects of Decyl-β-D-maltopyranoside on cellular barriers, this study offers insights into drug delivery systems and how compounds can be designed to improve absorption and efficacy (Marušić et al., 2013).

  • Characterization and two-dimensional crystallization of membrane component AlkB of the medium-chain alkane hydroxylase system from Pseudomonas putida GPo1.: This paper uses Decyl-β-D-maltopyranoside to stabilize and study membrane proteins crucial for bioremediation, offering advancements in environmental biotechnology and a deeper understanding of bacterial metabolism (Alonso & Roujeinikova, 2012).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service