The melting temperature range of this product is 62-68°C. See the link below to review the OmniPur Agarose products brochure:
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/340/931/10001515-021047-agarose-products-ms.pdf
2070-OP
Agarose
OmniPur® Grade, Low Melting
Synonym(s):
OmniPur® Agarose, Low Melting, 3,6-Anhydro-α-L-galacto-β-D-galactan
About This Item
Recommended Products
Quality Level
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
foreign activity
DNAse, none detected
Protease, none detected
RNAse, none detected
shipped in
ambient
SMILES string
O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O
InChI
1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1
InChI key
MJQHZNBUODTQTK-WKGBVCLCSA-N
Application
- Porous Agarose Layered Magnetic Graphene Oxide Nanocomposite for Virus RNA Monitoring in Wastewater.: Introduces a composite material incorporating agarose for enhanced viral RNA capture and detection in environmental samples, highlighting agarose′s utility in public health monitoring (He B et al., 2024).
Warning
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
what is the melting temperature that is not displayed
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service