Skip to Content
Merck
All Photos(1)

Key Documents

203788

Sigma-Aldrich

5-Bromo-4-chloro-3-indolylphosphate, Disodium Salt

Synonym(s):

5-Bromo-4-chloro-3-indolylphosphate, Disodium Salt, X-Phos, 2Na, BCIP, 2Na

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.47
Pricing and availability is not currently available.

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white to off-white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)([O-])([O-])Oc1c2c(ccc(c2Cl)Br)Nc1

InChI

1S/C8H6BrClNO4P/c9-4-1-2-5-7(8(4)10)6(3-11-5)15-16(12,13)14/h1-3,11H,(H2,12,13,14)/p-2

InChI key

QRXMUCSWCMTJGU-UHFFFAOYSA-L

Related Categories

General description

Substrate for visualizing alkaline phosphatase, including histochemical applications. When used together with nitroblue tetrazolium (NBT; Cat. No. 484235), produces purple insoluble end product that is read visually.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Horwitz, J.P., et al. 1966. J. Med. Chem.9, 447.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service