1.93260
Potassium chromate
for analysis EMPARTA® ACS
Synonym(s):
Potassium chromate
About This Item
Recommended Products
grade
ACS reagent
storage temp.
room temp
SMILES string
[Cr](=O)(=O)([O-])[O-].[K+].[K+]
InChI
1S/Cr.2K.4O/q;2*+1;;;2*-1
InChI key
XMXNVYPJWBTAHN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Inorganics and solvents for basic and standard applications manufactured locally.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service