Skip to Content
Merck
All Photos(1)

Key Documents

1.93103

Supelco

Potassium hydroxide pellets

for analysis EMPARTA® ACS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352305

grade

ACS reagent

form

pellets

pH

(alkaline)

solubility

water: soluble

anion traces

chloride (Cl-): 0.010%
phosphate, phosphite (as PO43-): 0.0005%
sulfate (SO42-): 0.003%

cation traces

Ca: 0.0050%
Fe: 0.010%
Mg: 0.0020%
N: 0.001%
Na: 0.20%
Ni: 0.0010%
heavy metals (as Ag): 0.001%

storage temp.

20-25°C

SMILES string

[OH-].[K+]

InChI

InChI=1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Packaging

1.93103.0521: 500 kg, Minimum order quantity: 1
1.93103.5021: 5 kg, Minimum order quantity: 1
1.93103.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service