Skip to Content
Merck
All Photos(1)

Key Documents

1.46379

Millipore

Tryptic Soy Broth Ready-to-use Bottles

bottle capacity 1000 mL, bottle filling volume 1000 mL, closure type, blue screw cap with 3 loci, box of 6 bottles

Synonym(s):

CASO, Casein Soybean Digest Broth, SCDB, TSB, Tryptic Soy Broth, Tryptone Soya Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

EP 2.6.1
JP 4.06
USP 72

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type blue screw cap with 3 loci
ready-to-use

packaging

box of 6 bottles

bottle capacity

1000 mL

bottle filling volume

1000 mL

pH

7.3

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Tryptic Soy Broth is a universal complex medium for the isolation and cultivation of fastidious aerobic bacteria, yeasts, and molds. The two highly nutritious peptones support the growth of a wide variety of aerobic and facultative anaerobic microorganisms, including fungi.

Application

Tryptic Soy Broth is used for the microbiological examination of non-sterile products. It is suitable for the sterility testing of substances, preparations, and products as well as a pre-enrichment broth for the test for specified microorganisms or total viable aerobic count determinations using the MPN method.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service