Skip to Content
Merck
All Photos(1)

Key Documents

124005

Sigma-Aldrich

Akt Inhibitor

The Akt Inhibitor controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Akt Inhibitor, 1L6-Hydroxymethyl-chiro-inositol-2-(R)-2-O-methyl-3-O-octadecyl- sn-glycerocarbonate, 1L6-Hydroxymethyl-chiro-inositol-2-(R)-2-O-methyl-3-O-octadecyl-sn-glycerocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H58O10
Molecular Weight:
578.78
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell permeable, reversible, and substrate competitive phosphatidylinositol ether analog that potently and selectively inhibits Akt (PKB) (IC50 of 5.0 µM). Moderately inhibits PI 3-K activity (IC50 = 83.0 µM). Also inhibits the growth of various cancer cell lines with IC50 values in the 1 - 10 µM range.

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes
Target IC50: 5.0 µM for Akt (PKB)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 weeks at -20°C.
Use anhydrous DMSO for reconstitution.

Other Notes

Hu, Y., et al. 2000 J. Med. Chem.43, 3045.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thanakorn Pungsrinont et al.
International journal of molecular sciences, 22(20) (2021-10-24)
Androgen deprivation therapy (ADT) and androgen receptor (AR)-targeted therapy are the gold standard options for treating prostate cancer (PCa). These are initially effective, as localized and the early stage of metastatic disease are androgen- and castration-sensitive. The tumor strongly relies
Sharanya Nag et al.
iScience, 26(4), 106307-106307 (2023-03-28)
Post-transcriptional regulation by RNA-binding proteins (RBPs) is a major mode of controlling gene expression under stress conditions. The RBP HuR regulates the translation/turnover of multiple mRNAs in stress responses. HuR is degraded in response to heat stress consequent to ubiquitination
Anke Dienelt et al.
Birth defects research, 114(16), 1056-1074 (2022-09-28)
Diabetes, which is characterized by an increase in blood glucose concentration, is accompanied by low bone turnover, increased fracture risk, and the formation of embryonic skeletal malformations. Yet, there are few studies elucidating the underlying alterations in signaling pathways leading
Wangqin Shu et al.
Molecular and cellular biology, 40(3) (2019-11-13)
Chronic hepatitis B (CHB) remains a global health problem, carrying a high risk for progression into cirrhosis and liver failure. Molecular chaperones are involved in diverse pathophysiological processes including viral infection. However, the role of molecular chaperones in hepatitis B
Ho Yong Kim et al.
Drug delivery, 29(1), 2897-2911 (2022-09-08)
Exosomes are nanosized extracellular vesicles secreted by various cell types, including those of the immune system, such as natural killer (NK) cells. They play a role in intercellular communication by transporting signal molecules between the cells. Recent studies have reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service