Skip to Content
Merck
All Photos(1)

Key Documents

1.16975

Supelco

Peracetic Acid Test Strips, reflectometric

1.0-22.5 mg/L (peracetic acid), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Peracetic Acid chemical test strips, Peracetic Acid test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Peracetic Acid Test, reflectometric, 1.0-22.5 mg/L (peracetic acid), for use with REFLECTOQUANT®

description

quantitative

Quality Level

product line

Reflectoquant®

specific analyte(s)

peracetic acid

reaction suitability

reaction type: Redox Reactions

measuring range

1.0-22.5 mg/L (peracetic acid)

compatibility

RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 1.0 - 22.5 mg/l Reflectoquant®
Peracetic acid is a frequently used disinfectant. The Reflectoquant Peracetic acid test strips are suited for the selective determination of the peracetic acid concentration in aqueous solutions and to check for the absence of peracetic acid after rinsing processes, also in cases in which hydrogen peroxide is present. Peracetic acid reacts with an aromatic amine to form a blue dye that is determined reflectometrically on the RQflex® 20. For pre-testing a sample in an economic and quick way to estimate the peracetic acid concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for peracetic acid.

Application


  • Efficient activation of peracetic acid by cobalt modified nitrogen-doped carbon nanotubes for drugs degradation: This study explores a novel method to enhance the efficacy of peracetic acid in degrading pharmaceutical compounds, highlighting its potential in environmental disinfection monitoring and safety applications in pharmaceutical manufacturing (Yao et al., 2024).

  • Ecotoxicological evaluation of effluent from bovine slaughterhouses disinfected by peracetic acid (PAA): This research assesses the environmental impact of peracetic acid used in the disinfection of wastewater from bovine slaughterhouses, using the bioindicator Girardia tigrina, which is crucial for environmental safety assessments (Pereira et al., 2024).

  • Peracetic acid activated by nickel cobaltite as effective oxidizing agent for BPA and its analogues degradation: This article presents a method for activating peracetic acid with nickel cobaltite to degrade BPA and its analogs effectively, addressing concerns over biocide efficacy and environmental safety (Kiejza et al., 2024).

  • Accelerated removal of naproxen in the iron-based peracetic acid activation system by chloride ions: The study focuses on enhancing reactive oxidative species in water treatment processes, showing how chloride ions can boost peracetic acid′s efficiency, important for biocide applications and safety testing (Yu et al., 2024).

Linkage

Replaces: 16975-1; 16975

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

See All

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service