Skip to Content
Merck
All Photos(1)

Key Documents

1.11755

Millipore

GranuCult® PALCAM agar (Base)

pkg of 500 g, according to FDA-BAM, ISO 11290, for Listeria spp.

Synonym(s):

Listeria selective agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

ISO 11290
according to FDA-BAM

Quality Level

sterility

non-sterile

form

powder

packaging

pkg of 500 g

manufacturer/tradename

NutriSelect® Prime

technique(s)

microbiological culture: suitable

pH

6.9-7.1 (25 °C, 68.8 g/L in H2O, after autoclaving)

solubility

68.8 g/L

bulk density

650 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Listeria spp.

General description

GranuCult® PALCAM agar (Base) provides quantitative cultivation of Listeria monocytogenes and inhibits the accompanying Gram-negative and most of the Gram-positive bacteria. Polymyxin, acriflavin, ceftazidime, and lithium chloride result in the selectivity of the medium. L. monocytogenes break down the esculin in the medium to glucose and esculetin. Esculetin forms an olive-green to black complex with iron (III) ions, which stains the colonies of L. monocytogenes. Mannitol-positive accompanying bacteria such as Staphylococci grow as yellow colonies if they are not inhibited.

Application

GranuCult® PALCAM agar (Base) is suitable for the detection and isolation of Listeria monocytogenes from feces, biological samples, foodstuffs, and heavily contaminated material from the environment.

Analysis Note

Base tested with PALCAM-Listeria selective supplement acc. to VAN NETTEN et.al. art. no. 1.12122.
Appearance (clearness): clear to slightly opalescent
Appearance (color): red
pH-value (25 °C): 7.0 - 7.4
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Inoculum on reference medium (Listeria monocytogenes ATCC 35152 (WDCM 00109)):
Inoculum on reference medium (Listeria monocytogenes ATCC 13932 (WDCM 00021)):
Inoculum on reference medium (Listeria innocua ATCC 33090 (WDCM 00017)):
Colony count (Listeria monocytogenes ATCC 35152 (WDCM 00109)):
Colony count (Listeria monocytogenes ATCC 13932 (WDCM 00021)):
Colony count (Listeria innocua ATCC 33090 (WDCM 00017)):
Recovery on test medium (Listeria monocytogenes ATCC 35152 (WDCM 00109)): ≥ 50 %
Recovery on test medium (Listeria monocytogenes ATCC 13932 (WDCM 00021)): ≥ 50 %
Recovery on test medium (Listeria innocua ATCC 33090 (WDCM 00017)): ≥ 50 %
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): total inhibition
Growth(Escherichia coli ATCC 8739 (WDCM 00012)): total inhibition
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): partial inhibition
Black zone (Listeria monocytogenes ATCC 35152 (WDCM 00109)): black zone
Black zone (Listeria monocytogenes ATCC 13932 (WDCM 00021)): black zone
Black zone (Listeria innocua ATCC 33090 (WDCM 00017)): black zone
Incubation: up to 48 hours at 35 ± 1 °C, aerobic
A recovery rate of 50 % is equivalent to a productivity value of 0.5.
The indicated colony counts result from the sum of a triple determination.
Reference media: Tryptic Soy Agar

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service