Skip to Content
Merck
All Photos(1)

Key Documents

1.10316

Millipore

Fractogel® EMD TMAE Hicap (M)

Synonym(s):

Biochromatography, Fractogel® Ion exchangers, Ion exchanger for LC (Fractogel®TSK/EMD), Fractogel® EMD TMAE Hicap (M)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

ligand

(Trimethylammoniumethyl)

Quality Level

description

strong anion exchanger, suspension in 20% ethanol and 150 mM NaCl (40-90 µm)

sterility

sterile (Caustic Stable)

form

resin

specific analyte(s)

MAbs

parameter

180 cm/hr flow rate
8 bar max. pressure

matrix active group

methacrylate

mean particle size

40-90 μm

capacity

180 mg binding capacity (BSA/mg of resin)

transition temp

flash point 35 °C (calculated)

density

1.430 g/cm3 at 20 °C

bulk density

1000 kg/m3

application(s)

recombinant protein

separation technique

anion exchange
strong anion exchange

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Strong anion exchange Fractogel® resin featuring the tentacle technology with a trimethylammoniumethyl functional group for purification of acidic and neutral proteins and peptides from multiple sources, DNA and virus removal, plasmid purification, vaccine purification, and more

Features and Benefits

Fractogel® EMD TMAE Hicap (M) enables users to:

  • Efficiently capture target protein and remove viruses, DNA, and endotoxins with its tentacle technology resulting in better production yields and high throughput
  • Reduce operating costs and resin replacement with enduring durability - allowing for multiple cycles of column regeneration and sanitization
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution


Due to the titration behavior of Fractogel® EMD TMAE Hicap (M) the ion exchange capacity can be used from pH 2 to 12. The separation of proteins is based on reversible electrostatic interactions between the negatively charged regions of the proteins′ surface and the support. Proteins are retained efficiently on Fractogel® EMD TMAE Hicap (M) when the pH of the buffer is about 1 unit above their isoelectric points (pl).

The strength of the binding depends on the following:
  • the buffer system
  • pH value of the buffer which determines the surface charge of the protein
  • the concentration of the counter ions
  • the charge density on the support (protein binding capacity)

Packaging

  • 1.10316.0100: Fractogel® EMD TMAE Hicap (M) Resin 100ml
  • 1.10316.0010: Fractogel® EMD TMAE Hicap (M) Resin 10ml
  • 1.10316.0500: Fractogel® EMD TMAE Hicap (M) Resin 500ml
  • 1.10316.5000: Fractogel® EMD TMAE Hicap (M) Resin 5L

Plastic bottle

Analysis Note

Appearance: Milky turbid suspension, free from impurities (foreign particles)
Microscopic evaluation: Uniform spherical particles,no agglomerates,no fines
Extractable matter (water): ≤ 0.03 %
Cerium: ≤ 1 µg/g
Pressure drop(column: ID=1.6 cm, L=10 cm at 5 ml/min): ≤ 1 bar
Particle size (d10): 37 - 45 µm
Particle size (d50): 48 - 60 µm
Particle size (d90): 63 - 77 µm
Colony forming units (TAMC + TYMC): ≤ 100 CFU/ml
Endotoxins: ≤ 1.00 EU/ml
Protein binding capacity (bovine serum albumin): 160 - 200 mg/ml
Functional test (b:a): ≤ 0.25
Functional test: Separation of conalbumin and human serum albumin

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service