Skip to Content
Merck
All Photos(4)

Key Documents

1.10020

Supelco

Nitrate Test Strips, colorimetric

greener alternative

10-500 mg/L (NO₃⁻), MQuant®, for use with MQuant® StripScan App

Synonym(s):

Nitrate chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Nitrate Test, colorimetric, 10-500 mg/L (NO3-), for use with MQuant® StripScan App

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

nitrate

reaction suitability

reaction type: Griess reaction

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

10-500 mg/L (NO3-)

compatibility

for use with MQuant® StripScan App

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric, with test strips, 10 - 25 - 50 - 100 - 250 - 500 mg/L (NO3-), MQuant®

The MQuant® Nitrate test strips can be used for quick and easy monitoring of drinking water, process water and wastewater, as well as in aquarium water where limits can quickly be exceeded. They can also be used to determine nitrate in food e.g. potatoes. On the strip nitrate ions are reduced to nitrite ions by a reducing agent, which then react to a red-violet dye. The nitrate concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. Each strip also features a second reaction zone (alert zone), the color of which changes in the presence of nitrite ions. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the nitrate concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® Nitrate test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Linkage

Replaces: 10020-1; 10020

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Reflectometric determination of nitrate in wastewater using Griess reagent and Reflectoquant® test strips.

Reflectometric determination of nitrate in wastewater using Griess reagent and Reflectoquant® test strips.

Reflectometric determination of nitrate in wastewater using Griess reagent and Reflectoquant® test strips.

Reflectometric determination of nitrate in wastewater using Griess reagent and Reflectoquant® test strips.

Related Content

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service