1.09613
Toluene-4-sulfonic acid monohydrate
for analysis EMSURE® ACS
Synonym(s):
Toluene-4-sulfonic acid monohydrate, p-Toluenesulfonic acid, Tosylic acid, Tosic acid, PTSA
About This Item
Recommended Products
grade
ACS reagent
Quality Level
product line
EMSURE®
Assay
≥99.0% anhydrous basis (acidimetric)
form
crystals
autoignition temp.
600 °C
potency
2570 mg/kg LD50, oral (Rat)
composition
Water, 9.5-11.5% Karl Fischer
impurities
≤0.1% Sulfated ash
pH
1 (20 °C, 650 g/L in H2O, anhydrous substance)
bp
140 °C/27 hPa
mp
56 °C (Elimination of water of crystallisation)
transition temp
flash point 180 °C
solubility
750 g/L
bulk density
510 kg/m3
anion traces
sulfate (SO42-): ≤0.3%
cation traces
Fe: ≤0.01%
Na: ≤0.002%
heavy metals (as Pb): ≤0.001%
storage temp.
15-25°C
SMILES string
[S](=O)(=O)([O-])c1ccc(cc1)C.[O+H3]
InChI
1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2
InChI key
KJIFKLIQANRMOU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Linkage
Analysis Note
Identity: passes test
Appearance of solution: conforms
Sulphate (SO₄): ≤ 0.3 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.01 %
Na (Sodium): ≤ 0.002 %
TLC-Test: passes test
Sulfated ash: ≤ 0.1 %
Water (according to Karl Fischer): 9.5 - 11.5 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1C - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service