Skip to Content
Merck
All Photos(1)

Key Documents

1.08298

Sigma-Aldrich

Xylol-IVD for Microscopy

for histology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.21
application(s):
diagnostic assay manufacturing
hematology
histology
bp:
136-145 °C/1013 hPa
vapor pressure:
8 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

8 hPa ( 20 °C)

Quality Level

form

liquid

IVD

for in vitro diagnostic use

color

colorless

kinematic viscosity

<0.9 cSt(20 °C)

bp

136-145 °C/1013 hPa

mp

-25 °C

transition temp

flash point 24 °C

solubility

0.175 g/L

density

0.87 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Xylene (isomeric mixture) - for histology, belongs to the aromatic hydrocarbons and can be used for for histological, cytological, and bacteriological investigation of sample material of human origin. Xylene is the gold standard and most common aromatic solvent which has been used for decades in routine histoprocessing (process for dehydrating the tissue) deparaffination, and clarification applications. A mixture of the ortho-, meta- and para- isomers provides for a higher degree of hydrophobicity prior to mounting and coverslipping. Histological grade Xylene was developed to be a robust clearing agent in both histological and cytological applications. It is a critical raw material in all histological staining procedures. It supports the QA process in Pathology. It is a CE-certified, IVD product. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Histological and cytological grade solvent for tissue processing, deparaffinization, and clearing.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service