Skip to Content
Merck
All Photos(6)

Key Documents

1.08114

Supelco

Tetrahydrofuran

≥99% (GC), EMPLURA®

Synonym(s):

THF, Tetramethylene oxide, Oxolane, Butylene oxide, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-726-8
NACRES:
NA.21
Assay:
≥99% (GC)
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Tetrahydrofuran, EMPLURA®

vapor density

2.5 (vs air)

Quality Level

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

impurities

≤0.002% Free acid (as CH3COOH)
≤0.01% Peroxide (as H2O2)
≤0.1% Water

refractive index

n20/D 1.407 (lit.)

pH

7-8 (20 °C, 200 g/L in H2O)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

transition temp

flash point -21.5 °C

density

0.889 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Self-Assembly of Polyoxometalate-Based Nanoparticle Surfactants in Solutions: Highlights the use of Tetrahydrofuran as a critical solvent in the synthesis and self-assembly of polyoxometalate-based nanoparticles, exploring applications that could impact multiple fields including polymer research, drug delivery systems, and nanostructured materials (Xia et al., 2024).

  • Highly sensitive fluorescence turn-OFF and reversible chemical sensor for Hg(2+) ion based on pyrene appended 2-thiohydantoin: Tetrahydrofuran is used as a key solvent in the development of a novel fluorescence-based chemical sensor for mercury detection, demonstrating its application in environmental monitoring and safety in laboratory settings (Gawas et al., 2024).

  • Effect of Plasma pretreatment and Graphene oxide ratios on the transport properties of PVA/PVP membranes for fuel cells: This research employs Tetrahydrofuran as a processing solvent in the creation of polyvinyl alcohol/polyvinylpyrrolidone membranes enhanced with graphene oxide, focusing on improvements in ion transport for fuel cell applications (Abdel-Hamed et al., 2024).

  • Optimized Incorporation of Furan into Diketopyrrolopyrrole-Based Conjugated Polymers for Organic Field-Effect Transistors: Utilizes Tetrahydrofuran as a solvent to aid in the synthesis of novel conjugated polymers, potentially leading to advancements in organic electronics and polymer research (Neu et al., 2024).

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as CH₃COOH): ≤ 0.002 %
Density (d 20 °C/ 4 °C): 0.887 - 0.889
Peroxide (as H₂O₂): ≤ 0.01 %
Water: ≤ 0.1 %
Stabilized with 2,6-Di-tert-butyl-4-methylphenol (BHT)

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service