Skip to Content
Merck

1.05999

SAFC

Parteck® Mn SD

EMPROVE® ESSENTIAL, Ph. Eur., USP, FCC

Pharma Manufacturing

Synonym(s):

Manganese(II) sulfate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MnO4S · xH2O
CAS Number:
Molecular Weight:
151.00 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-089-9

Agency

Ph. Eur.
USP

Quality Level

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

fine powder

potency

2150 mg/kg LD50, oral (Rat)

pH

3.0-3.5 (20 °C, 50 g/L in H2O)

mp

>449 °C (anhydrous substance)

density

2.95 g/cm3 at 20 °C

application(s)

pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[Mn+2].[S](=O)(=O)([O-])[O-].O

InChI

1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

ISPYRSDWRDQNSW-UHFFFAOYSA-L

General description

With formulation challenges in mind, we have used particle engineering technologies to develop functional excipients specifically for solid oral dosage forms. The products in our Parteck® portfolio feature unique particle properties tailored for excellent performance in tableting processes, for specific drug delivery technologies, or for solubility enhancement.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Parteck® Mn SD manganese sulfate is particle-engineered to a fine particle size and a narrow and reproducible particle size distribution (PSD), supporting a fine and even distribution. Its large particle surface area contributes to fast and quantitative dissolution, one of the key factors for good absorption and, consequently, good bioavailability.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service