Skip to Content
Merck
All Photos(1)

Key Documents

1.05084

Supelco

Potassium permanganate

for analysis (max. 0.000005% Hg) EMSURE® ACS

Synonym(s):

Potassium permanganate, Permanganic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
MDL number:
UNSPSC Code:
12352312
EC Index Number:
231-760-3
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
64 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor pressure

<0.01 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.0% (manganometric)

form

solid

potency

750 mg/kg LD50, oral (Rat)

impurities

≤0.1% Insoluble matter

pH

7-9 (20 °C, 20 g/L in H2O)

mp

>240 °C (decomposition)

solubility

64 g/L

density

2.70 g/cm3 at 20 °C

bulk density

1300‑1600 kg/m3

anion traces

chloride, chlorate (as Cl-): ≤0.005%
sulfate (SO42-): ≤0.01%

cation traces

Cu: ≤0.001%
Fe: ≤0.002%
Hg: ≤0.000005%
N: ≤0.003%
Pb: ≤0.002%

storage temp.

2-30°C

SMILES string

[Mn](=O)(=O)(=O)[O-].[K+]

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Related Categories

Application

  • Experimental investigation on strengthening of Zea mays root fibres for biodegradable composite materials using potassium permanganate treatment.: This research focuses on the treatment of Zea mays root fibers with potassium permanganate to enhance their mechanical properties for use in biodegradable composites, highlighting the role of chemical treatments in improving the performance of natural fibers in sustainable material applications (Blanco D et al., 2024).
  • Potential in-vitro antioxidant and anti-inflammatory activity of Martynia annua extract mediated Phytosynthesis of MnO(2) nanoparticles.: Explores the synthesis of manganese dioxide nanoparticles using Martynia annua extract, where potassium permanganate serves as an oxidizing agent to enhance the antioxidant and anti-inflammatory properties of the nanoparticles, showcasing its utility in medicinal chemistry (Perumal K et al., 2024).
  • Development of a thin layer chromatography-based method to detect sorbitol presence in milk and its applicability in formalin preserved milk samples.: Describes the development of a novel analytical method using thin layer chromatography and potassium permanganate to detect sorbitol in milk samples, demonstrating the adaptability of potassium permanganate in food safety and quality assurance processes (Sharma V et al., 2024).
  • Examining the physico-chemical, structural and thermo-mechanical properties of naturally occurring Acacia pennata fibres treated with KMnO(4).: Investigates the impact of potassium permanganate treatment on the properties of Acacia pennata fibers, focusing on their potential application in environmentally friendly composites, highlighting the use of potassium permanganate in modifying natural materials for enhanced utility (Flores ES et al., 2023).
  • Understanding the Risks of Diffusion of Cyanobacteria Toxins in Rivers, Lakes, and Potable Water.: Discusses the challenges of controlling cyanobacteria toxins in water bodies and the role of potassium permanganate as a potent oxidizing agent to mitigate these risks, emphasizing its importance in environmental protection and water treatment processes (Karaman R et al., 2023).

Analysis Note

Assay (manganometric): ≥ 99.0 %
Insoluble matter: ≤ 0.1 %
Chloride, Chlorate (as Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.01 %
Total nitrogen (N): ≤ 0.003 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.002 %
Hg (Mercury): ≤ 0.000005 %
Pb (Lead): ≤ 0.002 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service