Skip to Content
Merck
All Photos(1)

Key Documents

1.04617

Supelco

Hydroxylammonium sulfate

GR for analysis

Synonym(s):

Hydroxylammonium sulfate, Hydroxylamine sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HONH3)2SO4
CAS Number:
Molecular Weight:
164.14
MDL number:
UNSPSC Code:
12352301
EC Index Number:
233-118-8
NACRES:
NA.21
form:
solid

Quality Level

form

solid

potency

842 mg/kg LD50, oral (Rat)
1500-2000 mg/kg LD50, skin (Rabbit)

pH

3.6 (20 °C, 10 g/L in H2O)

mp

170 °C (decomposition)

solubility

587 g/L

density

1.88 g/cm3 at 20 °C

bulk density

1100 kg/m3

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])[O-].[N+H3]O.[N+H3]O

InChI

1S/2H4NO.H2O4S/c2*1-2;1-5(2,3)4/h2*2H,1H3;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

VGYYSIDKAKXZEE-UHFFFAOYSA-L

Analysis Note

Assay: ≥ 99 %
Chloride (Cl): ≤ 0.001 %
As (Arsenic): ≤ 0.00005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
Hg (Mercury): ≤ 0.000001 %
Pb (Lead): ≤ 0.0005 %
Sulfated ash (residue on ignition): ≤ 0.1 %

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service