Skip to Content
Merck
All Photos(1)

Documents

1.04106

Supelco

Glycolic acid

for analysis EMSURE® Reag. Ph Eur

Synonym(s):

Glycolic acid, Hydroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O3
CAS Number:
Molecular Weight:
76.05
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-180-5
NACRES:
NA.21

vapor pressure

10.8 hPa ( 80 °C)

Quality Level

product line

EMSURE®

Assay

≥99.5% (acidimetric)

form

solid

potency

2040 mg/kg LD50, oral (Rat)

impurities

≤0.5% Water (Karl Fischer)

pH

2 (20 °C, 50 g/L in H2O)

bp

100 °C (decomposition)

mp

74-78 °C

transition temp

flash point >300 °C (decomposition)

solubility

water: soluble at 20 °C

density

1.49 g/cm3 at 25 °C

bulk density

600 kg/m3

cation traces

Fe: ≤5 ppm
heavy metals (as Pb): ≤10 ppm

storage temp.

2-30°C

InChI

1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)

InChI key

AEMRFAOFKBGASW-UHFFFAOYSA-N

Application

  • An Overview of Poly(lactic-co-glycolic) Acid (PLGA)-Based Biomaterials for Bone Tissue Engineering: Discusses the synthesis and physicochemical characteristics of PLGA composites containing glycolic acid (P Gentile, V Chiono, I Carmagnola - International Journal of Molecular Sciences, 2014). Read more.

Analysis Note

Assay (acidimetric): ≥ 99.5 %
Identity (IR): passes test
Melting range (lower value): ≥ 75 °C
Melting range (upper value): ≤ 79 °C
Heavy metals (as Pb): ≤ 10 ppm
Iron (Fe): ≤ 5 ppm
Water (K. F.): ≤ 0.50 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

572.0 °F - (decomposition)

Flash Point(C)

> 300 °C - (decomposition)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service