Skip to Content
Merck
All Photos(1)

Key Documents

07-810

Sigma-Aldrich

Anti-phospho-Met (Tyr1230/Tyr1234/Tyr1235) Antibody

Upstate®, from rabbit

Synonym(s):

c-Met, cmet, HGFR, RCCP2, met proto-oncogene (hepatocyte growth factor receptor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
IF
WB
species reactivity:
mouse, human
technique(s):
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable
citations:
9

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr1230/pTyr1234/pTyr1235)

Gene Information

human ... MET(4233)
mouse ... Met(17295)
rat ... Met(24553)

Specificity

Recognizes Met phosphorylated on both Tyr1234/Tyr1235 and Tyr1230/Tyr1234/Tyr1235.

Immunogen

Peptide corresponding to amino acid region encompassing the human, mouse, and rat phospho-c-Met (Tyr1230/Tyr1234/Tyr1235)

Application

Anti-phospho-Met (Tyr1230/Tyr1234/Tyr1235) Antibody is an antibody against phospho-Met (Tyr1230/Tyr1234/Tyr1235) for use in IC, IF & WB.
Immunoblot Analysis: A 1:1000 dilution of this lot detected phospho-Met (Tyr1230/Tyr1234/Tyr1235) in RIPA lysate of HGF treated 293T cells.

Quality

Routinely evaluated by immunoblot.

Target description

155 kDa

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federica Chiara et al.
The Journal of biological chemistry, 278(31), 29352-29358 (2003-05-15)
The met oncogene, encoding the high affinity hepatocyte growth factor receptor, is the only known gene inherited in human cancer that is invariably associated with somatic duplication of the mutant locus. Intriguingly, mutated Met requires ligand stimulation in order to
P Longati et al.
Oncogene, 9(1), 49-57 (1994-01-01)
The tyrosine kinase encoded by the MET proto-oncogene (p190MET) is the receptor for Hepatocyte Growth Factor/Scatter Factor (HGF/SF). Previous work has shown that autophosphorylation of p190MET enhances its enzymatic activity and that the major phosphorylation site is Tyr1235, located in
Kwang-Hoon Song et al.
Hepatology (Baltimore, Md.), 46(6), 1993-2002 (2007-10-10)
Bile acid synthesis in the liver is regulated by the rate-limiting enzyme cholesterol 7alpha-hydroxylase (CYP7A1). Transcription of the CYP7A1 gene is inhibited by bile acids and cytokines. The rate of bile acid synthesis is reduced immediately after partial hepatectomy and
Xiaobin Wang et al.
Journal of cellular and molecular medicine, 25(21), 10164-10174 (2021-10-16)
Mantle cell lymphoma (MCL) is a haematologic malignancy. The proteasome inhibitor (PI) bortezomib has been approved to treat MCL, but resistance has emerged through mechanisms that remain unclear. This study aimed to explore the mechanism of PI resistance in MCL
c-Met modulates RPE migratory response to laser-induced retinal injury.
Kasaoka, M; Ma, J; Lashkari, K
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service