Skip to Content
Merck
All Photos(1)

Key Documents

06-236

Sigma-Aldrich

Anti-Giα1/2 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
IP
WB
species reactivity:
mouse, human, rat, bovine
technique(s):
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
8

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat, bovine

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GNAI1(2770)

Specificity

Recognizes Giα1 and Giα2.

Immunogen

10 residue synthetic peptide (KNNLKDCGLF) corresponding to the c-terminus of Gia1 and Gia2

Application

Detect Giα1/2 using this Anti-Giα1/2 Antibody validated for use in IC, IP & WB.
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Quality

routinely evaluated by immunoblot on Rat Brain Microsomal Preparation or solubilized plasma membranes from mouse 3T3/A31 fibroblasts

Target description

40 kDa

Physical form

A diluted rabbit serum in PBS (50 mM KHPO4, 140 mM NaCl). Frozen solution
Format: Purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Widely expressed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Disulfide bonds in the extracellular calcium-polyvalent cation-sensing receptor correlate with dimer formation and its response to divalent cations in vitro
Ward, D. T., et al
The Journal of Biological Chemistry, 273, 14476-14483 (1998)
P Barrett et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 14(11), 1619-1628 (2000-08-06)
Mimicking short photoperiod melatonin signals (16 h exposure) on primary cell cultures of melatonin target cells of the ovine pars tuberalis (PT) results in an enhanced cAMP response to forskolin stimulation relative to untreated cells, a phenomenon termed sensitization. The
J M Sands et al.
The Journal of clinical investigation, 99(6), 1399-1405 (1997-03-15)
During antidiuresis, increases in vasopressin (AVP)-elicited osmotic water permeability in the terminal inner medullary collecting duct (tIMCD) raise luminal calcium concentrations to levels (> or = 5 mM) above those associated with the formation of calcium-containing precipitates in the urine.
A Abbaci et al.
Oncogene, 37(6), 756-767 (2017-10-24)
B-cell chronic lymphocytic leukemia (B-CLL) cells are resistant to apoptosis, and consequently accumulate to the detriment of normal B cells and patient immunity. Because current therapies fail to eradicate these apoptosis-resistant cells, it is essential to identify alternative survival pathways
C J Lynch et al.
The Journal of clinical investigation, 83(6), 2050-2062 (1989-06-01)
Adenylate cyclase in liver plasma membranes from streptozotocin-diabetic (STZ) or BB/Wor spontaneously diabetic rats showed increased responsiveness to GTP, glucagon, fluoroaluminate, and cholera toxin. Basal or forskolin-stimulated activity was unchanged in STZ rats, but increased in BB/Wor rats. No change

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service