Skip to Content
Merck
All Photos(1)

Key Documents

05-22-2500

Sigma-Aldrich

Formyl-Met-Leu-Phe-OH

Chemotactic peptide for polymorphonuclear leukocytes.

Synonym(s):

Formyl-Met-Leu-Phe-OH, fMLP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31N3O5S
CAS Number:
Molecular Weight:
437.55
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.28

Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to off-white

solubility

DMSO: 100 mg/mL
methanol: 20 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C21H31N3O5S/c1-14(2)11-17(23-19(26)16(22-13-25)9-10-30-3)20(27)24-18(21(28)29)12-15-7-5-4-6-8-15/h4-8,13-14,16-18H,9-12H2,1-3H3,(H,22,25)(H,23,26)(H,24,27)(H,28,29)/t16-,17-,18-/m0/s1

InChI key

PRQROPMIIGLWRP-BZSNNMDCSA-N

General description

Chemotactic peptide for polymorphonuclear leukocytes. Induces superoxide release from neutrophils.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Chemotactic peptide for polymorphonuclear leukocytes
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

fMet-Leu-Phe-OH

Other Notes

Bautista, A.P., and Elliott, K.E. 1994. Life Sci. 54, 721.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service