Skip to Content
Merck
All Photos(1)

Key Documents

Z-003

Supelco

Zopiclone solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClN6O3
CAS Number:
Molecular Weight:
388.81
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[NH+]1CCN(CC1)C(=O)OC2N(C(=O)c3nccnc23)c4ccc(Cl)cn4

InChI

1S/C17H17ClN6O3/c1-22-6-8-23(9-7-22)17(26)27-16-14-13(19-4-5-20-14)15(25)24(16)12-3-2-11(18)10-21-12/h2-5,10,16H,6-9H2,1H3/p+1

InChI key

GBBSUAFBMRNDJC-UHFFFAOYSA-O

General description

A certified solution standard suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, pharmaceutical research, or urine drug testing.  Zopiclone is a non-benzodiazepine hypnotic sold under the trade names Imovane® and ZImovane® and is prescribed for the treatment of insomnia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Imovane is a registered trademark of Sanofi-Aventis
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coma after single dose of zopiclone.
Annelie K Hintzen et al.
The American journal on addictions, 21(3), 291-291 (2012-04-13)
To breathe, perchance not to wake?
David M Rapoport
Journal of applied physiology (Bethesda, Md. : 1985), 112(2), 247-248 (2011-10-29)
Nico J Diederich et al.
Journal of the neurological sciences, 314(1-2), 12-19 (2011-11-29)
Sleep symptoms in Parkinson's disease (PD) are frequent and have multifactorial and multilayered causes. Primary involvement of sleep/wake regulating centers in the brainstem, sleep problems caused by the nocturnal manifestation of motor and dysautonomic signs and medication-induced sleep problems are
Milena Araújo Tonon et al.
Analytical and bioanalytical chemistry, 405(1), 267-273 (2012-11-07)
A new high-performance liquid chromatographic method with triple quadrupole mass spectrometry detection was developed and validated for the quantification of zopiclone enantiomers in rat brain samples. Zopiclone enantiomers were resolved on a CHIRALPAK AD column with a mobile phase consisting
Alan Wayne Jones et al.
Forensic science international, 222(1-3), 118-123 (2012-06-08)
The concentrations of zolpidem and zopiclone were determined in peripheral blood samples in two forensic materials collected over a 10-year period (2001-2010). The z-hypnotics were determined in venous blood from living subjects (impaired drivers) and in femoral blood from deceased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service