Skip to Content
Merck
All Photos(1)

Key Documents

N-102

Supelco

Norfluoxetine-D6 oxalate solution

1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®

Synonym(s):

Deuterated norfluoxetine oxalate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10D6F3NO · C2H2O4
Molecular Weight:
391.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

FC(F)(F)c1ccc(cc1)OC(CCN)(c2c(c(c(c(c2[2H])[2H])[2H])[2H])[2H])[2H].OC(=O)C(=O)O

InChI

1S/C16H16F3NO.C2H2O4/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;3-1(4)2(5)6/h1-9,15H,10-11,20H2;(H,3,4)(H,5,6)/i1D,2D,3D,4D,5D,15D;

InChI key

FDWJCEPFCGIDPF-OYCZTGALSA-N

General description

Norfluoxetine is a primary metabolite of the selective serotonin reuptake inhibitor (SSRI) fluoxetine, an antidepressant. This internal standard is suitable for quantitation of norfluoxetine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis.

Application


  • Advanced Material Characterization: Norfluoxetine-D₆ oxalate is utilized in cutting-edge research involving synchrotron radiation for high-resolution mapping on carbonate stones. This application highlights its use in sophisticated material sciences, helping to understand mineral treatments and interactions at the microscale (Massinelli et al., 2024).


  • Phytoextraction Enhancement in Agriculture: While primarily used in pharmaceutical contexts, related research utilizes the principles underlying the actions of compounds like Norfluoxetine-D₆ oxalate to enhance the phytoextraction potential of plants in contaminated environments. This research extends to the utilization of nitrogen fertilizers and organic acids to remediate polluted dredged sediment (Stojanov et al., 2024).


  • Pesticide Application in Agriculture: Similar to its application in pharmaceuticals, the principles of effective compound utilization as seen in Norfluoxetine-D₆ oxalate are applied in agriculture to understand the infection and control mechanisms of Sclerotium rolfsii in crops like peppers. This helps in the effective management of stem rot diseases using targeted pesticide treatments (Song et al., 2024).


  • Environmental Science Research: Research into enhanced rock weathering as a method for carbon dioxide removal showcases the indirect applications of analytical techniques similar to those used in studying Norfluoxetine-D₆ oxalate. This study evaluates the potential in tropical environments like Costa Rica to combat climate change through geochemical processes (Ryan et al., 2024).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongsheng Peng et al.
The Journal of biological chemistry, 294(21), 8529-8542 (2019-04-11)
The farnesoid X receptor (FXR) is a member of the nuclear receptor superfamily of bile acid-activated transcription factors and an important regulator of cell proliferation, apoptosis, and Wnt signaling. Down-regulated expression of FXR plays an important role in some malignancies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service