Skip to Content
Merck
All Photos(1)

Key Documents

890870P

Avanti

16:0 TAP

Avanti Research - A Croda Brand 890870P, powder

Synonym(s):

1,2-dipalmitoyl-3-trimethylammonium-propane (chloride salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H76NO4Cl
CAS Number:
Molecular Weight:
646.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 200 mg (890870P-200mg)
pkg of 1 × 25 mg (890870P-25mg)
pkg of 1 × 500 mg (890870P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890870P

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Cl-]

InChI

1S/C38H76NO4.ClH/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-37(40)42-35-36(34-39(3,4)5)43-38(41)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2;/h36H,6-35H2,1-5H3;1H/q+1;/p-1

InChI key

KYARZEFZZOGPGS-UHFFFAOYSA-M

General description

16:0 TAP is a cationic lipid.

Application

16:0 TAP is suitable for the preparation of giant unilamellar vesicles (GUVs) by dissolving in chloroform:methanol and added into cell membrane to simulate the charged groups in the cell membrane.

Packaging

20 mL Clear Glass Screw Cap Vial (890870P-500mg)
5 mL Amber Glass Screw Cap Vial (890870P-200mg)
5 mL Amber Glass Screw Cap Vial (890870P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dispersion of atmospheric fine particulate matters in simulated lung fluid and their effects on model cell membranes
Zhou Q, et al.
The Science of the Total Environment, 542, 36-43 (2016)
Role of bovine serum albumin and humic acid in the interaction between SiO2 nanoparticles and model cell membranes
Wei X, et al.
Environmental Pollution (Barking, Essex : 1987), 219, 1-8 (2016)
Chun Liufu et al.
International journal of nanomedicine, 14, 8923-8941 (2019-12-10)
Cancer stem cells (CSCs) are responsible for cancer therapeutic resistance and metastasis. To date, in addition to surgery, chemotherapy, and radiotherapy, gene delivery has emerged as a potential therapeutic modality for ovarian cancer. Efficient and safe targeted gene delivery is
Bao Zhu et al.
Chemosphere, 247, 125876-125876 (2020-01-25)
Aluminium oxide nanoparticles (Al2O3 NPs) potentially cause health hazards after their release into the environment. The crystalline phase of Al2O3 NPs determines their surface structure and the number of functional groups. The adsorption of natural organic matter (NOM) or biomolecules
Ling Liu et al.
The Science of the total environment, 734, 139229-139229 (2020-05-26)
The induced membrane damage is a key mechanism for the cytotoxicity of graphene nanosheets (GNSs). In this research, the physical interaction of GNSs on model membranes was investigated using artificial membranes and plasma membrane vesicles. The effects of the GNSs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service