Skip to Content
Merck
All Photos(1)

Key Documents

870273P

Avanti

18:1 Biotinyl Cap PE

Avanti Research - A Croda Brand 870273P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(cap biotinyl) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H102N4O11PNaS
CAS Number:
Molecular Weight:
1105.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 2 × 100 mg (870273P-200mg)
pkg of 1 × 25 mg (870273P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870273P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCCCCNC(CCCC[C@](SC[C@@]1(NC(N2)=O)[H])([C@]12[H])[H])=O)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C57H103N4O11PS.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33-41-54(64)69-46-49(72-55(65)42-34-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)47-71-73(67,68)70-45-44-59-53(63)39-32-31-37-43-58-52(62)40-36-35-38-51-56-50(48-74-51)60-57(66)61-56;/h17-20,49-51,56H,3-16,21-48H2,1-2H3,(H,58,62)(H,59,63)(H,67,68)(H2,60,61,66);/q;+1/p-1/b19-17-,20-18-;/t49-,50+,51+,56+;/m1./s1

InChI key

VQSGNVZURNOPBT-XEAQQBNDSA-M

General description

18:1 Biotinyl Cap PE, also known as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(cap biotinyl) is a head group modified lipid containing biotinyl cap PE.

Application

18:1 Biotinyl Cap PE may be used in giant unilamellar vesicles (GUVs) and black lipid membranes (BLMs) preparation.

Packaging

5 mL Clear Glass Sealed Ampule (870273P-200mg)
5 mL Clear Glass Sealed Ampule (870273P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David K Y Zhang et al.
Nature protocols, 15(3), 773-798 (2020-01-15)
Synthetic antigen-presenting cells (APCs) are used to mediate scalable ex vivo T-cell expansion for adoptive cell therapy. Recently, we developed APC-mimetic scaffolds (APC-ms), which present signals to T cells in a physiological manner to mediate rapid and controlled T-cell expansion.
Véronique Yvette Ntsogo Enguéné et al.
Frontiers in microbiology, 6, 541-541 (2015-06-18)
Efflux pumps are membrane transporters that actively extrude various substrates, leading to multidrug resistance (MDR). In this study, we have designed a new test that allows investigating the assembly of the MexA-MexB-OprM efflux pump from the Gram negative bacteria Pseudomonas
Lucrecia Alberdi et al.
Journal of cell science, 133(9) (2020-05-16)
Salmonella enterica is an intracellular bacterial pathogen. The formation of its replication niche, which is composed of a vacuole associated with a network of membrane tubules, depends on the secretion of a set of bacterial effector proteins whose activities deeply
Florian Schlosser et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 370(1661), 20140028-20140028 (2014-12-24)
Cells are sensitive to mechanical cues from their environment and at the same time generate and transmit forces to their surroundings. To test quantitatively forces generated by cells not attached to a substrate, we used a dual optical trap to
Yanqi Yu et al.
ACS nano (2018-11-14)
Understanding the binding of nanoparticles to receptors on biomembranes is critical to the development and screening of therapeutic materials. A prevailing understanding is that multivalent ligand-receptor binding leads to slower and confined translational motion of nanoparticles. In contrast, we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service